α-Naphtholphthalein

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.06246.0001 1.06246.0005
1.06246.0001EA 4970 NOK
1.06246.0001 1.06246.0005
α-Naphtholphthalein
α-Naphtholphthalein
Formel: C₂₈H₁₈O₄
MDL nummer: MFCD00036202
CAS-nummer: 596-01-0
EINECS: 209-875-5

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) practically insoluble
Melting Point 253 - 255 °C
Molar Mass 418.45 g/mol
Bulk Density 350 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR