Alizarin red S (sodium salt)

Leverandør: Merck
Warning

Synonymer: C.I. 58005

M-Clarity™ quality level = MQ200

1.06278.0025 1.06278.0100
1.06278.0025EA 1270 NOK
1.06278.0025 1.06278.0100
Alizarin red S (sodium salt)
Alizarin red S (sodium salt)
Formel: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Molar Mass 342.26 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR