Natriumhydrogenkarbonat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ300

1.06329.0500 1.06329.1000 1.06329.5000 1.06329.9050 1.06329.9025
1.06329.0500EA 437 NOK
1.06329.0500 1.06329.1000 1.06329.5000 1.06329.9050 1.06329.9025
Natriumhydrogenkarbonat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Natriumhydrogenkarbonat
Formel: NaHCO₃
Smeltepunkt: 270 °C
Tetthet: 2,159 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003528
CAS-nummer: 144-55-8
EINECS: 205-633-8
Merck Indeks: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 96 g/l (20 °C)
Melting Point 270 °C (decomposition)
Molar Mass 84.01 g/mol
Bulk Density 1000 kg/m3
Vapor Pressure 8.3 hPa (30 °C)
Density 2.22 g/cm3 (20 °C)
pH 8.6 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR