Natriumhydrogensulfat monohydrat

Leverandør: Merck
Danger

Synonymer: Sodium bisulphate monohydrate

1.06350.9025
1.06350.9025EA 0 NOK
1.06350.9025
Natriumhydrogensulfat monohydrat
Natriumhydrogensulfat monohydrat
Formel: NaHSO₄·H₂O
MW: 138.08 g/mol
Smeltepunkt: 183
Tetthet: 2.1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149210
CAS-nummer: 10034-88-5
EINECS: 231-665-7
FN: 3260
Merck Indeks: 13,08658

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR