Natriumkarbonat, vannfri, EMSURE® ISO for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.06392.0500 1.06392.1000 1.06392.5000 1.06392.9025
1.06392.0500EA 736 NOK
1.06392.0500 1.06392.1000 1.06392.5000 1.06392.9025
Natriumkarbonat, vannfri, EMSURE® ISO for analysis, Supelco®
Natriumkarbonat
Formel: Na₂CO₃
Kokepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 854 °C
Tetthet: 2,53 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003494
CAS-nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Indeks: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 220 g/l (20 °C)
Melting Point 854 °C
Molar Mass 105.99 g/mol
Bulk Density 1100 kg/m3
Boiling Point 1600 °C (decomposition)
Density 2.53 g/cm3 (20 °C)
pH 11.5 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR