Natriumklorid, Suprapur® for sporanalyse, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.06406.0500 1.06406.0050 1.06406.0250
1.06406.0500EA 8710 NOK
1.06406.0500 1.06406.0050 1.06406.0250
Natriumklorid, Suprapur® for sporanalyse, Supelco®
Natriumklorid
Formel: NaCl
Kokepunkt: 1413 °C (1013 hPa)
Smeltepunkt: 801 °C
Tetthet: 2,16 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3
Merck Indeks: 14,08599

Bestill nå

Spesifikasjon Testresultater

Solubility 358 g/l (20 °C)
Melting Point 801 °C
Molar Mass 58.44 g/mol
Bulk Density 1140 kg/m3
Boiling Point 1461 °C (1013 hPa)
Vapor Pressure 1.3 hPa (865 °C)
Density 2.17 g/cm3 (20 °C)
pH 6 - 9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR