Natriumtiosulfat pentahydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ300

1.06516.0500 1.06516.1000 1.06516.5000 1.06516.9025
1.06516.0500EA 451 NOK
1.06516.0500 1.06516.1000 1.06516.5000 1.06516.9025
Natriumtiosulfat pentahydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Natriumtiosulfat pentahydrat
Formel: Na₂S₂O₃.5H₂O
Smeltepunkt: 48 °C
Tetthet: 1,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149186
CAS-nummer: 10102-17-7
EINECS: 231-867-5
Merck Indeks: 12,08844

Bestill nå

Spesifikasjon Testresultater

Solubility 701 g/l (20 °C)
Melting Point 48 °C
Molar Mass 248.21 g/mol
Bulk Density 1000 kg/m3
Density 1.74 g/cm3 (20 °C)
pH 6.0 - 7.5 (100 g/l, H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR