di-Natriumdisulfitt, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.06528.0100 1.06528.0500 1.06528.1000 1.06528.5000 1.06528.9050
1.06528.0100EA 232 NOK
1.06528.0100 1.06528.0500 1.06528.1000 1.06528.5000 1.06528.9050
di-Natriumdisulfitt, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
di-Natriumdisulfitt
Formel: Na₂S₂O₅
Smeltepunkt: 150 °C
Tetthet: 1,48 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167602
CAS-nummer: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Indeks: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 650 g/l (20 °C)
Melting Point 150 °C (decomposition)
Molar Mass 190.11 g/mol
Bulk Density 1000 - 1200 kg/m3
Density 2.36 g/cm3 (20 °C)
pH 4.0 - 4.8 (50 g/l, H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR