Sodium polyphosphate (Graham's salt), EMPLURA®, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.06529.1000 1.06529.5000 1.06529.9050
1.06529.1000EA 571 NOK
1.06529.1000 1.06529.5000 1.06529.9050
Sodium polyphosphate (Graham's salt), EMPLURA®, Supelco®
Sodium polyphosphate (Graham's salt)
Formel: (NaO₃P)n
Tetthet: 2,4 g/cm³ (20 °C)
MDL nummer: MFCD00134118
CAS-nummer: 50813-16-6
EINECS: 233-782-9
Merck Indeks: 12,08814

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR