Natriumperborat tetrahydrat, pure, Supelco®

Leverandør: Merck
Danger

Synonymer: Sodium peroxometaborate tetrahydrate

M-Clarity™ quality level = MQ200

1.06560.9050
1.06560.9050EA 0 NOK
1.06560.9050
Natriumperborat tetrahydrat, pure, Supelco®
Natriumperborat tetrahydrat
Formel: NaH₂BO₄·4H₂O
Smeltepunkt: 60 °C
Tetthet: 1,73 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149231
CAS-nummer: 10486-00-7
EINECS: 239-172-9

Bestill nå

Spesifikasjon Testresultater

Solubility 23 g/l (20 °C)
Melting Point 60 °C (decomposition)
Molar Mass 153.88 g/mol
Bulk Density 800 kg/m3
Density 1.73 g/cm3
pH 10 (15 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR