Natriumsalisylat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.06601.0250 1.06601.1000 1.06601.2500
1.06601.0250EA 450 NOK
1.06601.0250 1.06601.1000 1.06601.2500
Natriumsalisylat, EMSURE® for analysis, Supelco®
Natriumsalisylat
Formel: HOC₆H₄COONa
Smeltepunkt: 200 °C
Tetthet: 0,32 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002440
CAS-nummer: 54-21-7
EINECS: 200-198-0
Merck Indeks: 12,08819

Bestill nå

Spesifikasjon Testresultater

Solubility 1000 g/l (20 °C)
Melting Point 200 °C
Molar Mass 160.10 g/mol
Bulk Density 320 kg/m3
Density 0.32 g/cm3 (20 °C)
pH 6.5 (100 g/l, H2O, 20 °C)
Ignition Point 250 °C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR