Natriumsulfat, EMSURE® ACS for analysis, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ300

1.06637.1000 1.06637.0500 1.06637.9025
1.06637.1000EA 737 NOK
1.06637.1000 1.06637.0500 1.06637.9025
Natriumsulfat, EMSURE® ACS for analysis, Supelco®
Sodium sulphate
Formel: Na₂SO₄
Kokepunkt: 1429 °C (1013 hPa)
Smeltepunkt: 888 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
EINECS: 231-820-9
Merck Indeks: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 44.55 g/l (20 °C)
Melting Point 888 °C
Molar Mass 142.04 g/mol
Bulk Density 1400 - 1600 kg/m3
Density 2.70 g/cm3 (20 °C)
pH 4 - 7 (200 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR