Nickel (II) klorid heksahydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.06717.0250 1.06717.1000
1.06717.0250EA 1170 NOK
1.06717.0250 1.06717.1000
Nickel (II) klorid heksahydrat, EMSURE® for analysis, Supelco®
Nickel (II) klorid heksahydrat
Formel: NiCl₂·6H₂O
Smeltepunkt: 140 °C
Tetthet: 1,84 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149809
CAS-nummer: 7791-20-0
EINECS: 231-743-0
FN: 3288
ADR: 6.1,III
Merck Indeks: 13,06529

Bestill nå

Spesifikasjon Testresultater

Solubility 553 g/l (20 °C)
Melting Point 1001 °C (anhydrous substance),decomposes
Molar Mass 237.66 g/mol
Bulk Density 640 kg/m3
Boiling Point 987 °C (sublimed),(anhydrous substance)
pH 4.9 (100 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR