Nikkel(II)nitrat heksahydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.06721.0100 1.06721.0250 1.06721.1000
1.06721.0100EA 783 NOK
1.06721.0100 1.06721.0250 1.06721.1000
Nikkel(II)nitrat heksahydrat, EMSURE® for analysis, Supelco®
Nikkel(II)nitrat heksahydrat
Formel: Ni(NO₃)₂·6H₂O
Kokepunkt: 137 °C (1013 hPa)
Tetthet: 2,05 g/cm³ (20 °C)
MDL nummer: MFCD00149805
CAS-nummer: 13478-00-7
EINECS: 236-068-5
FN: 2725
ADR: 5.1,III
Merck Indeks: 12,06596

Bestill nå

Spesifikasjon Testresultater

Solubility 940 g/l (20 °C)
Melting Point 56.7 °C
Molar Mass 290.81 g/mol
Bulk Density 800 kg/m3
Boiling Point 136.7 °C (1013 hPa)
Density 2.05 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR