3-Nitrophenol, Millipore®

Leverandør: Merck
Warning

Synonymer: m-Nitrophenol

M-Clarity™ quality level = MQ200

1.06794.0025
1.06794.0025EA 2270 NOK
1.06794.0025
3-Nitrophenol, Millipore®
3-Nitrophenol
Formel: C₆H₅NO₃
Kokepunkt: 271 °C (1013 hPa)
Smeltepunkt: 94…95 °C
Tetthet: 1,485 g/cm³ (20 °C)
Flash punkt: >100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007240
CAS-nummer: 554-84-7
EINECS: 209-073-5
FN: 1663
ADR: 6.1,III
Merck Indeks: 13,06652

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 95 - 98 °C
Molar Mass 139.11 g/mol
Bulk Density 640 kg/m3
Boiling Point 194 °C (93 hPa)
Flash Point 100 °C
Density 1.49 g/cm3 (20 °C)
pH 6.6 - 8.6 (H2O, 20 °C)
Ignition Point 400 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR