4-Nitrofenol ≥99.5% indicator

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

1.06798.0025 1.06798.0100
1.06798.0025EA 1220 NOK
1.06798.0025 1.06798.0100
4-Nitrofenol ≥99.5% indicator
4-Nitrofenol
Formel: C₆H₅NO₃
Kokepunkt: 279 °C (1013 hPa)
Smeltepunkt: 110…114 °C
Tetthet: 1,48 g/cm³ (20 °C)
Flash punkt: 169 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007331
CAS-nummer: 100-02-7
EINECS: 202-811-7
FN: 1663
ADR: 6.1,III
Merck Indeks: 13,06654

Bestill nå

Spesifikasjon Testresultater

Hill Formula C₆H₅NO₃
Chemical formula O₂NC₆H₄OH
Molar Mass 139.11 g/mol
Boiling point 279 °C (1013 hPa) (decomposition)
Density 1.48 g/cm³ (20 °C)
Flash point 169 °C
Ignition temperature 510 °C
Melting Point 110 - 114 °C
pH value 4.4 (5 g/l, H₂O, 24 °C)
Bulk density 550 kg/m³
Solubility 16 g/l (25 °C)
LD 50 dermal LD50 Rat 1024 mg/kg
Storage class 6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR