Etanol absolute, EMPARTA® ACS analytical reagent, Supelco®

Leverandør: Merck
Danger

Synonymer: Ethyl alcohol

M-Clarity™ quality level = MQ200

1.07017.2511 1.07017.2511_T 1.07017.9026 1.07017.9026_T 1.07017.4000
1.07017.2511EA 0 NOK
1.07017.2511 MERC1.07017.2511_T 1.07017.9026 MERC1.07017.9026_T 1.07017.4000
Etanol absolute, EMPARTA® ACS analytical reagent, Supelco®
Etanol

Bio-Based Ethanol
Instead of synthetic ethanol, we use bioethanol produced from grain or sugar cane. High quality, affordability, and ready availability make our bioethanol an obvious choice for a sustainable future.


  • Produced from grain or sugar cane, a renewable source
  • Less toxic than synthetic ethanol (no toxic by-products)
  • Reliable availability
  • Production method is safer for the environment

Formel: C₂H₅OH
Kokepunkt: 78,3 °C (1013 hPa)
Smeltepunkt: –117 °C
Tetthet: 0.7895 g/cm³ (20 °C)
Flash punkt: 12 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
FN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Indeks: 12,03806

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) completely miscible
Melting Point -114.5 °C
Molar Mass 46.07 g/mol
Boiling Point 78.3 °C (1013 hPa)
Vapor Pressure 59 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.36
Explosion Limit 3.5 - 15 %(V)
Density 0.790 - 0.793 g/cm3 (20 °C)
pH 7.0 (10 g/l, H2O, 20 °C)
Ignition Point 425 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR