Dietyleter Stabilised, EMPARTA® ACS analysis_for

Leverandør: Merck
Danger

Synonymer: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisering: EtOH
MQ200

1.07062.4000
1.07062.4000EA 2600 NOK
1.07062.4000
Dietyleter Stabilised, EMPARTA® ACS analysis_for
Dietyleter
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kokepunkt: 35
Smeltepunkt: -123
Flash punkt: -40
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011646
CAS-nummer: 60-29-7
FN: 1155
ADR: 3 I
Merck Indeks: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR