Fenolftalein 0,375% in methanol

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.07238.0250 1.07238.1000
1.07238.0250EA 739 NOK
1.07238.0250 1.07238.1000
Fenolftalein 0,375% in methanol
Fenolftalein
Formel: C₂₀H₁₄O₄
Tetthet: 0,8 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005913
CAS-nummer: 77-09-8
EINECS: 201-004-7
FN: 1230
ADR: 3 (6.1),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Flash Point 11 °C
Explosion Limit 5.5 - 36.5 %(V)
Density 0.80 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR