Bly (II) acetat trihydrat

Leverandør: Merck
Danger

Synonymer: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

1.07372.1000 1.07372.5000 1.07372.9050
1.07372.1000EA 740 NOK
1.07372.1000 1.07372.5000 1.07372.9050
Bly (II) acetat trihydrat
Bly (II) acetat trihydrat
Formel: (H₃CCOO)₂Pb·3H₂O
Kokepunkt: 280 °C (1013 hPa)
Smeltepunkt: 75 °C
Tetthet: 2,55 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150023
CAS-nummer: 6080-56-4
EINECS: 206-104-4
FN: 1616
ADR: 6.1,III
Merck Indeks: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR