Bly(II)karbonat, EMSURE® ACS for analysis, Supelco®

Leverandør: Merck
Danger

Synonymer: Lead carbonat

M-Clarity™ quality level = MQ300

1.07381.0250
1.07381.0250EA 1110 NOK
1.07381.0250
Bly(II)karbonat, EMSURE® ACS for analysis, Supelco®
Bly(II)karbonat
Formel: PbCO₃
MW: 267.21 g/mol
Smeltepunkt: 315
Tetthet: 6.43 g/cm³ (20 °C)
MDL nummer: MFCD00064631
CAS-nummer: 598-63-0
EINECS: 209-943-4
FN: 2291
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) practically insoluble
Melting Point 315 °C (decomposition)
Molar Mass 267.21 g/mol
Bulk Density 1300 kg/m3
Density 6.6 g/cm3 (20 °C)
pH 5.8 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR