Bly(II)acetat basisk, anhydrous, EMSURE® for the analysis of sugar acc. to horne, Supelco®

Leverandør: Merck
Danger

Synonymer: Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate

M-Clarity™ quality level = MQ300

1.07414.1000 1.07414.9030
1.07414.1000EA 1090 NOK
1.07414.1000 1.07414.9030
Bly(II)acetat basisk, anhydrous, EMSURE® for the analysis of sugar acc. to horne, Supelco®
Bly(II)acetat basisk
Formel: (CH₃COO)₂Pb·2Pb(OH)₂
Smeltepunkt: 198…205 °C
Tetthet: 3,58 g/cm3 (22 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011154
CAS-nummer: 51404-69-4
EINECS: 215-630-3
FN: 2291
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Assay (PbO) ≥ 33.0 %
In water insoluble matter ≤ 1.0 %
Substances insoluble in dilute acetic acid ≤ 0.02 %
Chloride (Cl) ≤ 0.003 %
Nitrate und Nitrite (as NO₃) passes test
Ca (Calcium) ≤ 0.01 %
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.002 %
K (Potassium) ≤ 0.02 %
Na (Sodium) ≤ 0.05 %
Loss on Drying (105 °C) ≤ 1.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR