4-(2-Pyridylazo)resorcinol monosodim salt monohydrate, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.07533.0005
1.07533.0005EA 3470 NOK
1.07533.0005
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate, Supelco®
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
Formel: C₁₁H₈N₃NaO₂·1H₂O
MDL nummer: MFCD00150712
CAS-nummer: 16593-81-0
EINECS: 236-339-8

Bestill nå

Spesifikasjon Testresultater

Solubility 38 g/l (20 °C)
Molar Mass 255.21 g/mol
Bulk Density 630 kg/m3
pH 9.8 (10 g/l, H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR