Sjøsand for analysis, renset med syre, calcinated, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.07712.1000 1.07712.5000 1.07712.9025 1.07712.9010
1.07712.1000EA 1850 NOK
1.07712.1000 1.07712.5000 1.07712.9025 1.07712.9010
Sjøsand for analysis, renset med syre, calcinated, Supelco®
Sjøsand
Formel: SiO₂
Kokepunkt: >2200 °C (1013 hPa)
Smeltepunkt: 1713 °C
Tetthet: 2,65 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
CAS-nummer: 14808-60-7
EINECS: 231-545-4

Bestill nå

Spesifikasjon Testresultater

Substances soluble in hydrochloric acid ≤ 0.2 %
Chloride (Cl) ≤ 0.005 %
Acid-soluble Iron (Fe) ≤ 0.01 %
Loss on ignition (900 °C) ≤ 0.05 %
Particle size (0.1 - 0.315 mm) ≥ 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR