Tinn, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.07807.0250
1.07807.0250EA 0 NOK
1.07807.0250
Tinn, Supelco®
Tinn
Formel: Sn
Kokepunkt: 2270 °C (1013 hPa)
Smeltepunkt: 232 °C
Tetthet: 7,31 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Indeks: 12,09587

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 232 °C
Molar Mass 118.71 g/mol
Boiling Point 2260 °C (1013 hPa)
Density 7.3 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR