Tinn(II)klorid dihydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.07815.0100 1.07815.0250 1.07815.1000 1.07815.9025
1.07815.0100EA 1220 NOK
1.07815.0100 1.07815.0250 1.07815.1000 1.07815.9025
Tinn(II)klorid dihydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Tinn(II)klorid dihydrat
Formel: SnCl₂·2H₂O
Kokepunkt: 623 °C (1013 hPa)
Smeltepunkt: 246 °C
Tetthet: 2,71 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149863
CAS-nummer: 10025-69-1
EINECS: 231-868-0
FN: 3260
ADR: 8,II
Merck Indeks: 13,08861

Bestill nå

Spesifikasjon Testresultater

Solubility 1187 g/l (20 °C)
Melting Point 246 °C (anhydrous substance)
Molar Mass 225.63 g/mol
Bulk Density 1250 kg/m3
Boiling Point 623 °C (1013 hPa) (anhydrous substance)
Density 2.71 g/cm3 (20 °C)
pH 1 - 2 (100 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR