Tiourea, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Thiocarbamide, Sulphourea

MQ200

1.07978.9025
1.07978.9025EA 0 NOK
1.07978.9025
Tiourea, Sigma-Aldrich®
Tiourea
Formel: NH₂CSNH₂
MW: 76.12 g/mol
Smeltepunkt: 174…177 °C
Tetthet: 1.435 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008067
CAS-nummer: 62-56-6
EINECS: 200-543-5
FN: 3077
ADR: 9 III
Merck Indeks: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR