Tetrahydrofuran, anhydrous (max. 0.005% H₂O) for DNA and peptides synthesis

Leverandør: Merck
Danger

Synonymer: Butylene oxide, THF, Diethylene oxide

M-Clarity™ quality level = MQ200

1.08107.1000
1.08107.1000EA 1975 NOK
1.08107.1000
Tetrahydrofuran, anhydrous (max. 0.005% H₂O) for DNA and peptides synthesis
Tetrahydrofuran

These reagents are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency.

Formel: C₄H₈O
Kokepunkt: 66 °C (1013 hPa)
Tetthet: 0,888 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005356
CAS-nummer: 109-99-9
EINECS: 203-726-8
FN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Indeks: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C
Vapor Pressure 173 hPa (20 °C)
Flash Point -21.5 °C
Refractive Index 1.407 (20 °C)
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm3 (20 °C)
pH 7 - 8 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Saturation concentration 557 g/m3 (20 °C) Air
Evaporation number 2.3
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR