Triethanolamine hydrochloride, Millipore®

Supplier: Merck


M-Clarity™ quality level = MQ100

1.08357.0100 1.08357.1000
1.08357.0100EA 535 NOK
1.08357.0100 1.08357.1000
Triethanolamine hydrochloride, Millipore®
Triethanolamine hydrochloride
Formula: C₆H₁₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00012596
CAS Number: 637-39-8
EINECS: 211-284-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR