Triethanolamine hydrochloride, Millipore®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.08357.0100 1.08357.1000
1.08357.0100EA 535 NOK
1.08357.0100 1.08357.1000
Triethanolamine hydrochloride, Millipore®
Triethanolamine hydrochloride
Formel: C₆H₁₆ClNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012596
CAS-nummer: 637-39-8
EINECS: 211-284-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR