Titriplex® solution A for the determination of alkaline earth in water (hardness), Titrisol®, Supelco®
Leverandør: Merck
M-Clarity™ quality level = MQ100
1.08419.1000
1.08419.1000EA 819 NOK
1.08419.1000
Titriplex® solution A for the determination of alkaline earth in water (hardness), Titrisol®, Supelco®
Titriplex® solution A
for the determination of alkaline earth metals in water 1 ml ≙ 56 mg CaO/l using 100 ml of water
1 ml of reagent is equivalent to 5,6 degree German hardness when using 100 ml of water.
1°d ≙ 10 mg CaO/l
Bestill nå
Spesifikasjon Testresultater
Efficiency value 5.57 - 5.63 °dH
Determination method Complexometric titration
The efficiency value of this solution was determined with volumetric standard calcium carbonate (article number 1.02410). The expanded measurement uncertainty is ± 0.7 % (k=2 coverage factor for 95% coverage probability).
The certified value is traceable to primary standard NIST SRM 682 (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard calcium carbonate, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.