Titriplex® III concentrate 0.01 M, Titrisol®, Supelco®

Leverandør: Merck
Warning

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

MQ100

1.08446.0001
1.08446.0001EA 330.03 NOK
1.08446.0001
Titriplex® III concentrate 0.01 M, Titrisol®, Supelco®
EDTA dinatrium salt

Concentration after dilution to 1 liter: c(C₁₀H₁₄N₂Na₂O₈ ∙ 2 H₂O) = 0.01 mol/l


The concentration of this solution was determined with volumetric standard zinc (article number 1.02409). The determined titer at 20°C was 1.000 with an expanded measurement uncertainty of ± 0.004 (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 682 (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard zinc, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Amount-of-substance concentration 0.01000 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR