Zink stabilised, pulver, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

1.08774.1000 1.08774.9050
1.08774.1000EA 1210 NOK
1.08774.1000 1.08774.9050
Zink stabilised, pulver, Supelco®
Zinc
Formel: Zn
Kokepunkt: 907 °C (1013 hPa)
Tetthet: 7,13 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011291
CAS-nummer: 7440-66-6
EINECS: 231-175-3
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) (reaction),insoluble
Melting Point 420 °C
Molar Mass 65.39 g/mol
Bulk Density 1800 - 2700 kg/m3
Boiling Point 908 °C (1013 hPa)
Vapor Pressure 1.33 hPa (487 °C)
Density 7.14 g/cm3 (20 °C)
pH (H2O) not applicable

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR