Folin-Ciocalteu's reagent for analysis of phenols, Supelco®

Leverandør: Merck
Danger

Synonymer: Folin & Ciocalteu’s phenol reagent

1.09001.0100 1.09001.0500
1.09001.0100EA 455 NOK
1.09001.0100 1.09001.0500
Folin-Ciocalteu's reagent for analysis of phenols, Supelco®
Folin-Ciocalteu's reagent
Tetthet: 1.22 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00132625
FN: 3264
ADR: 8 III

Bestill nå

Spesifikasjon Testresultater

Equivalent acid c(H+)= 2 mol/l (2N)
Sensitivity to phenol Conforms
Sensitivity to bovine serum albumin Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR