Hoechst wax C, micropowder tableting aid for X-ray analysis, Supelco®

Leverandør: Merck

Synonymer: N,N'-1,2-Ethanediylbisoctadecanamide, Ethylene Distearamide, N,N'-Ethylenebisstearamide, Hoechst wax C, N,N'-Ethylenebisoctadecanamide

M-Clarity™ quality level = MQ100

1.09014.1000
1.09014.1000EA 518.68 NOK
1.09014.1000
Hoechst wax C, micropowder tableting aid for X-ray analysis, Supelco®
N,N'-Ethylenebis(stearamide)

Hoechst wax C micropowder tabletting aid for X-ray fluorescence analysis. CAS 110-30-5, pH (H₂O, 20 °C) Not applicable.

Formel: C₃₈H₇₆N₂O₂
MW: 593.04 g/mol
Smeltepunkt: 144 °C
Flash punkt: 270 °C (DIN 51758)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00059224
CAS-nummer: 110-30-5
EINECS: 203-755-6

Bestill nå

Spesifikasjon Testresultater

Bi (Bismuth) ≤ 10 µg/g
Co (Cobalt) ≤ 5 µg/g
Cr (Chromium) ≤ 5 µg/g
Cu (Copper) ≤ 5 µg/g
Fe (Iron) ≤ 5 µg/g
Mn (Manganese) ≤ 5 µg/g
Ni (Nickel) ≤ 5 µg/g
Pb (Lead) ≤ 10 µg/g
Sr (Strontium) ≤ 5 µg/g
Tl (Thallium) ≤ 10 µg/g
Zn (Zinc) ≤ 5 µg/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR