Schiff reagent for analysis of aldehydes, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.09034.1000
1.09034.1000EA 763 NOK
1.09034.1000
Schiff reagent for analysis of aldehydes, Supelco®
Schiff reagent

For the analytical determination of aldehydes in the ppm range.


For histology we recommend Schiff's reagent item 109033.

Bestill nå

Spesifikasjon Testresultater

Assay (SO₂) 0.15 - 0.30 %
Application test conforms
pH 1.5 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR