Sølvnitrat 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Leverandør: Merck
Danger

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

M-Clarity™ quality level = MQ100

1.09080.1000
1.09080.1000EA 2943.75 NOK
1.09080.1000
Sølvnitrat 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sølvnitrat
Formel: AgNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
FN: 1760
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR