Air samplers, MAS-100 Iso NT®
Leverandør: Merck
1.09168.0001 1.09173.0001 1.09174.0001
1.09168.0001EA 0 NOK
1.09168.0001 1.09173.0001 1.09174.0001
Air samplers, MAS-100 Iso NT®
Prøvetakere Luftprøvetakere
The MAS-100 Iso NT® is a high-performance instrument based on the impaction principle. The head (using standard 90 mm Petri dishes) can be installed at the critical control points with all electronic and moving parts kept outside the critical zone.
  • Integrated mass-flow sensor
  • Dialogue multilingual software
  • In-line sterilisation, 100 sample and alarm log
  • RABS-version without sterilisation cycle
  • Remote start and SQS mode − SeQuential Sampling over a period of time
  • Double valve and pump system for sterilisation of the head and tube in line with the isolator