Air sampler, MAS-100 NT®
Leverandør: Merck
1.09191.0001 1.17274.0001
1.09191.0001EA 0 NOK
1.09191.0001 1.17274.0001
Air sampler, MAS-100 NT®
Prøvetakere Luftprøvetakere
The perforated sampling head may be sterilised and can be adjusted to sample horizontally.
  • Petri dishes (90 mm) or contact plates (55 mm)
  • Programmable delay and individually pre-selected adjustable volumes
  • Up to 100 samples can be logged with date, time, user and volume
  • Fully automatic calibration cycle, automatic creation of certificate with digital anemometer DA-100 NT® and PC
  • Mass flow sensor to ensure a constant airflow rate
  • USB port to communicate with a computer
  • Autoclavable sampling head to avoid any contamination
Suitable for applicationsMore Product Information