Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l vandig løsning, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.09193.0100 1.09193.0500
1.09193.0100EA 835 NOK
1.09193.0100 1.09193.0500
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l vandig løsning, Supelco®
Ferroin (1,10-phenanthrolineferrous sulphate complex)
Formel: C₃₆H₂₄FeN₆SO₄
Tetthet: 1,01 g/cm³ (20 °C)
MDL nummer: MFCD00036428
CAS-nummer: 14634-91-4
EINECS: 238-676-6

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 2.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR