Alkali blue (C.I. 42765) indicator, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.09196.0025
1.09196.0025EA 909 NOK
1.09196.0025
Alkali blue (C.I. 42765) indicator, Supelco®
Alkali blue (C.I. 42765)
Formel: C₃₇H₂₉N₃O₃S
MDL nummer: MFCD00083221
CAS-nummer: 1324-76-1
EINECS: 215-385-2

Bestill nå

Spesifikasjon Testresultater

Molar Mass 595.73 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR