Krystall fiolett ethanolic aqueous solution primary stain for Gram staining, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: CI 42555

1.09218.0500 1.09218.2500
1.09218.0500EA 407 NOK
1.09218.0500 1.09218.2500
Krystall fiolett ethanolic aqueous solution primary stain for Gram staining, Sigma-Aldrich®
Krystall fiolett

The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative.


The mureine structure of the bacteria walls is the basis of the color affinity. Bacteria will be stained with Gram's crystal violet solution - an aniline dye - in the first step. After the treatment with iod solution (Lugol’s solution), a dye-iod complex will form. During the decolorizing step, this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized . Gram-negative bacteria will be counterstained by safranin solution and appear orange.

Formel: C₂₅H₃₀ClN₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011750
CAS-nummer: 548-62-9
FN: 1170
ADR: 3,III
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR