Kovac's indole reagent for microbiology pH <1 (H₂O, 20 °C), Millipore®

Leverandør: Merck
Danger

Synonymer: 4-(Dimethylamino)benzaldehyde solution

M-Clarity™ quality level = MQ100

1.09293.0100
1.09293.0100EA 372.6 NOK
1.09293.0100
Kovac's indole reagent for microbiology pH <1 (H₂O, 20 °C), Millipore®
Kovac's indole reagent

Contains 4-(Dimethylamino) benzaldehyde

Formel: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00003381
CAS-nummer: 100-10-7
FN: 2920
ADR: 8 (3) II

Bestill nå

Spesifikasjon Testresultater

Indole formation (Escherichia coli ATCC 25922 (WDCM 00013)) +
Indole formation (Proteus vulgaris ATCC 8427) +
Indole formation (Enterobacter aerogenes ATCC 13048) -
Tested in DEV-Tryptophan broth art. no. 1.10694. (after incubation 24 hrs.; 35 °C aerobic).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR