Kaliumhydroksid 0.5 mol/l (0.5 N) in methanol, Titripur® volumetrisk løsning, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.09351.1000 1.09351.1003
1.09351.1000EA 722 NOK
1.09351.1000 1.09351.1003
Kaliumhydroksid 0.5 mol/l (0.5 N) in methanol, Titripur® volumetrisk løsning, Supelco®
Kaliumhydroksid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: KOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
FN: 3286
ADR: 3 (6.1, 8),II

Bestill nå

Spesifikasjon Testresultater

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025
Measurement uncertainty +/- 0.0015
Traceability NIST SRM

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR