Buffer løsninger, klar til bruk, justert for 25 °C, Certipur®

Leverandør: Merck

1.09444.0500 1.09445.0500 1.09445.4000 1.99038.0500 1.99054.0500 1.99054.4000 1.09446.0500 1.99036.0500 1.09406.4000 1.09407.0500 1.09407.4000 1.09408.4000 1.09409.0500 1.09409.4000 1.09442.0500 1.09442.4000 1.09408.0500 1.09441.0500 1.99057.0500 1.99057.4000 1.99050.0500 1.99050.4000 1.09441.4000 1.99041.0500 1.99041.4000
1.09444.0500EA 464 NOK
1.09444.0500 1.09445.0500 1.09445.4000 1.99038.0500 1.99054.0500 1.99054.4000 1.09446.0500 1.99036.0500 1.09406.4000 1.09407.0500 1.09407.4000 1.09408.4000 1.09409.0500 1.09409.4000 1.09442.0500 1.09442.4000 1.09408.0500 1.09441.0500 1.99057.0500 1.99057.4000 1.99050.0500 1.99050.4000 1.09441.4000 1.99041.0500 1.99041.4000
Buffer løsninger, klar til bruk, justert for 25 °C, Certipur®
Buffere Buffere, pH

Materials for the precise calibration and monitoring of pH meters.


  • Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
  • Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
  • Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
  • Reference temperature: +25 °C


All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.


Forpakning: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)

Bestill nå

SPESIFIKASJONER

Environmentally Preferable Sustainable Packaging

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR