Testpapirer, Supelco®
Leverandør: Merck
1.09511.0003
1.09511.0003EA 623 NOK
1.09511.0003
Testpapirer, Supelco®
Testpapir Indikatorpapir
Lead (II) acetate paper is used for the determination of sulphide and hydrogen sulphide.