Starch potassium iodide paper Reag.Ph.Eur., Supelco®
Leverandør: Merck
1.09512.0003
1.09512.0003EA 395.36 NOK
1.09512.0003
Starch potassium iodide paper Reag.Ph.Eur., Supelco®
Testpapir Indikatorpapir
Potassium iodine paper is used for the determination of oxidising agents.
Limited Time Sale
Select items below are currently on sale