pH indicator papers, MQuant®, Supelco®
Leverandør: Merck
1.09527.0001 1.10232.0001 1.09526.0003 1.09486.0003 1.10962.0003 1.09489.0003 1.09521.0003 1.09555.0003 1.09557.0003 1.09514.0003 1.09556.0003
1.09527.0001EA 519.88 NOK
1.09527.0001 1.10232.0001 1.09526.0003 1.09486.0003 1.10962.0003 1.09489.0003 1.09521.0003 1.09555.0003 1.09557.0003 1.09514.0003 1.09556.0003
pH indicator papers, MQuant®, Supelco®
Testpapir pH-indikatorstrips
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips