2',7'-Dichlorofluorescein, Supelco®

Leverandør: Merck

Synonymer: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

M-Clarity™ quality level = MQ200

1.09676.0005
1.09676.0005EA 3825 NOK
1.09676.0005
2',7'-Dichlorofluorescein, Supelco®
2',7'-Dichlorofluorescein
Formel: C₂₀H₁₀Cl₂O₅
MDL nummer: MFCD00005047
CAS-nummer: 76-54-0
EINECS: 200-968-6

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Molar Mass 401.2 g/mol
Bulk Density 390 kg/m3

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR