Formamid, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.09684.1000 1.09684.2500
1.09684.1000EA 822 NOK
1.09684.1000 1.09684.2500
Formamid, EMSURE® for analysis, Supelco®
Formamid
Formel: HCONH₂
Kokepunkt: 103 °C (2 kPa)
Smeltepunkt: 2 °C
Tetthet: 1,1334 g/cm³ (20 °C)
Flash punkt: 79 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007941
CAS-nummer: 75-12-7
EINECS: 200-842-0
Merck Indeks: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (N; Kjeldahl) ≥ 99.2 %
Identity (IR) passes test
Appearance Clear, colorless liquid.
Colour (APHA) ≤ 10 APHA
Density (d 20 °C/ 4 °C) 1.132 - 1.135
Solidification point 2.0 - 3.0 °C
Boiling point (2 kPa) about 103
Chloride (Cl) ≤ 1 ppm
Cd (Cadmium) ≤ 1 ppm
Cu (Copper) ≤ 1 ppm
Fe (Iron) ≤ 1 ppm
Pb (Lead) ≤ 1 ppm
Zn (Zinc) ≤ 1 ppm
Formic acid ≤ 0.02 %
Ammonium Formate ≤ 0.1 %
Sulfated ash (600 °C) ≤ 0.005 %
Water (according to Karl Fischer) ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR